2-Bromo-4,6-xilidina ≥97.0% (by GC, titration analysis)

Proveedor: TCI
Atención

B1908-25G B1908-5G
TCIAB1908-25GEA 90 EUR
TCIAB1908-25G TCIAB1908-5G
2-Bromo-4,6-xilidina ≥97.0% (by GC, titration analysis)
2-Bromo-4,6-xilidina
Formula: C₈H₁₀BrN
Número MDL: MFCD00047826
Núm. CAS: 41825-73-4
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8a139817eaf200609e0bdd19acdbb479814aed2&vid=f8a139817eaf200609e0bdd19acdbb479814aed2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light orange to Dark red to Black powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 43.0 to 46.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR