(S)-(-)-3-tert-Butylamino-1,2-propanediol ≥96.0% (por GC)

Proveedor: TCI

B1987-5G
TCIAB1987-5GEA 78 EUR
TCIAB1987-5G
(S)-(-)-3-tert-Butylamino-1,2-propanediol ≥96.0% (por GC)
(S)-(-)-3-tert-Butylamino-1,2-propanediol
Formula: C₇H₁₇NO₂
Número MDL: MFCD00190165
Núm. CAS: 30315-46-9
EINECS: 250-125-1

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 96.0 %
Melting point 79.0 to 82.0 deg-C
Specific rotation [a]20/D -27.0 to -31.0 deg (C=2, 1mol/L HCl)
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR