N-(tert-Butoxycarbonyl)-D-tyrosine ≥98.0% (by titrimetric analysis)

Proveedor: TCI

B2963-1G B2963-5G
TCIAB2963-1GEA 41 EUR
TCIAB2963-1G TCIAB2963-5G
N-(tert-Butoxycarbonyl)-D-tyrosine ≥98.0% (by titrimetric analysis)
N-(tert-Butoxycarbonyl)-D-tyrosine
Formula: C₁₄H₁₉NO₅
Número MDL: MFCD00063030
Núm. CAS: 70642-86-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -1.0 to -4.0 deg (C=2, AcOH)

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR