1-Benzyl-L-aspartate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: L-Aspartic Acid 1-Benzyl Ester, H-Asp-OBzl

B2994-1G B2994-5G
TCIAB2994-1GEA 17 EUR
TCIAB2994-1G TCIAB2994-5G
1-Benzyl-L-aspartate ≥98.0% (by HPLC, titration analysis)
1-Benzyl-L-aspartate
Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
Número MDL: MFCD00063186
Núm. CAS: 7362-93-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR