1-Benzyl-D-glutamate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: D-Glutamic Acid 1-Benzyl Ester, H-D-Glu-OBzl

B2995-1G B2995-5G
TCIAB2995-1GEA 108 EUR
TCIAB2995-1G TCIAB2995-5G
1-Benzyl-D-glutamate ≥98.0% (by HPLC, titration analysis)
1-Benzyl-D-glutamate
Formula: C₁₂H₁₅NO₄
Número MDL: MFCD00237065
Núm. CAS: 79338-14-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d12f7eaf3ee512b4224374ca5d4b3a8195d7a7&vid=7dc8e6f918628193468e9183cbc9451138f597b2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -10.0 to -13.0 deg (C=1, 1mol/l HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR