5-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate hydrate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: N-Fmoc-L-glutamic acid 5-tert-butyl ester, Fmoc-Glu(OtBu)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic Acid 5-tert-Butyl Ester, 5-tert-Butyl N-Fmoc-L-glutamate

B3167-25G B3167-5G
TCIAB3167-25GEA 46 EUR
TCIAB3167-25G TCIAB3167-5G
5-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate hydrate ≥98.0% (by HPLC, titration analysis)
5-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate hydrate
Formula: C₂₄H₂₇NO₆
Número MDL: MFCD00037135
Núm. CAS: 71989-18-9
EINECS: 276-253-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Melting point 87.0 to 93.0 deg-C
Specific rotation [a]20/D -7.0 to -10.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR