2-Bromo-9,10-diphenylanthracene ≥95.0% (por GC)

Proveedor: TCI

B3442-1G
TCIAB3442-1GEA 783 EUR
TCIAB3442-1G
2-Bromo-9,10-diphenylanthracene ≥95.0% (por GC)
2-Bromo-9,10-diphenylanthracene
Formula: C₂₆H₁₇Br
MW: 409,33 g/mol
Número MDL: MFCD14708167
Núm. CAS: 201731-79-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 185.0 to 189.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR