4,4'-Bis(chloromethyl)-2,2'-bipyridyl ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: 4,4'-Bis(chloromethyl)-2,2'-bipyridine

B3572-1G
TCIAB3572-1GEA 621 EUR
TCIAB3572-1G
4,4'-Bis(chloromethyl)-2,2'-bipyridyl ≥98.0% (by titrimetric analysis)
4,4'-Bis(chloromethyl)-2,2'-bipyridyl
Formula: C₁₂H₁₀Cl₂N₂
MW: 253.13 g/mol
Número MDL: MFCD20265579
Núm. CAS: 138219-98-4
UN: 3261
ADR: 8 III

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR