5-Benzotriazolecarboxylic acid ≥96.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

B3869-5G B3869-25G
TCIAB3869-5GEA 41 EUR
TCIAB3869-5G TCIAB3869-25G
5-Benzotriazolecarboxylic acid ≥96.0% (by HPLC, titration analysis)
5-Benzotriazolecarboxylic acid
Formula: C₇H₅N₃O₂
Número MDL: MFCD00012318
Núm. CAS: 23814-12-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR