1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid ≥95.0% (by GC, titration analysis)

Proveedor: TCI
Atención

B3880-1G
TCIAB3880-1GEA 54 EUR
TCIAB3880-1G
1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid ≥95.0% (by GC, titration analysis)
1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid
Formula: C₁₀H₁₇NO₄
Temperatura de almacenaje: Nevera
Número MDL: MFCD02179181
Núm. CAS: 59378-75-5

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 133.0 to 139.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR