N,N',N''-Trimethyldiethylenetriamine ≥95.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: 2,2'-Bis(methylamino)-N-methyldiethylamine, N,N-Bis[2-(methylamino)ethyl]methylamine, N1,N2-Dimethyl-N1-(2-(methylamino)ethyl)ethane-1,2-diamine

B4304-5ML
TCIAB4304-5MLEA 144 EUR
TCIAB4304-5ML
N,N',N''-Trimethyldiethylenetriamine ≥95.0% (by GC, titration analysis)
N,N',N''-Trimethyldiethylenetriamine
Formula: C₇H₁₉N₃
MW: 145,25 g/mol
Pto. Ebullición: 56°C
Número MDL: MFCD00025637
Núm. CAS: 105-84-0
EINECS: 203-337-3
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f82ce1351a16ceec7017895c870f8330d7e6048&vid=4f82ce1351a16ceec7017895c870f8330d7e6048&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR