4-Bromo-1-naphthaleneboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: (4-Bromonaphthalen-1-yl)boronic acid

B4463-1G
TCIAB4463-1GEA 97 EUR
TCIAB4463-1G
4-Bromo-1-naphthaleneboronic acid (contains varying amounts of Anhydride)
4-Bromo-1-naphthaleneboronic acid
Formula: C₁₀H₈BBrO₂
MW: 250,88 g/mol
Número MDL: MFCD08701764
Núm. CAS: 145965-14-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 108.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR