trans,trans-1,5-Bis(4-methoxyphenyl)-1,4-pentadien-3-one ≥98.0% (por GC)

Proveedor: TCI

B4467-200MG B4467-1G
TCIAB4467-200MGEA 36 EUR
TCIAB4467-200MG TCIAB4467-1G
trans,trans-1,5-Bis(4-methoxyphenyl)-1,4-pentadien-3-one ≥98.0% (por GC)
trans,trans-1,5-Bis(4-methoxyphenyl)-1,4-pentadien-3-one
Formula: C₁₉H₁₈O₃
Número MDL: MFCD00014901
Núm. CAS: 37951-12-5

Order Now

Resultados test especificaciones

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR