Benzylamine hydroiodide, low water content ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

B4566-1G B4566-5G
TCIAB4566-1GEA 63 EUR
TCIAB4566-1G TCIAB4566-5G
Benzylamine hydroiodide, low water content ≥98.0% (by HPLC, total nitrogen)
Benzylamine hydroiodide
Formula: C₇H₉N
Núm. CAS: 45579-91-7

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 176.0 to 180.0 deg-C
Water max. 100 ppm

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR