Z-Glu-OtBu ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

B4912-5G B4912-1G
TCIAB4912-5GEA 74 EUR
TCIAB4912-5G TCIAB4912-1G
Z-Glu-OtBu ≥98.0% (by HPLC, titration analysis)
Z-Glu-OtBu
Formula: C₁₇H₂₃NO₆
Número MDL: MFCD00077016
Núm. CAS: 5891-45-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 81.0 to 85.0 deg-C
Specific rotation [a]20/D -24.0 to -28.0 deg (C=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR