3,3'-(Butane-1,4-diyl)bis(1-vinyl-3-imidazolium) bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC)

Proveedor: TCI
Atención

B5421-1G
TCIAB5421-1GEA 139 EUR
TCIAB5421-1G
3,3'-(Butane-1,4-diyl)bis(1-vinyl-3-imidazolium) bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC)
3,3'-(Butane-1,4-diyl)bis(1-vinyl-3-imidazolium) bis(trifluoromethanesulphonyl)imide
Formula: C₁₈H₂₀F₁₂N₆O₈S₄
Núm. CAS: 1312310-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7102e168be5ddc616ca5d1fe1c7d9e3400ce4cc2&vid=7102e168be5ddc616ca5d1fe1c7d9e3400ce4cc2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 96.0 %
Melting point 80.0 to 84.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR