tert-Butyl (2-hydroxyethyl)(methyl)carbamate ≥98.0% (por GC)

Proveedor: TCI
Atención

Sinónimos: N-(tert-Butoxycarbonyl)-N-methylethanolamine, N-Boc-N-methyl-2-aminoethanol, tert-Butyl (2-Hydroxyethyl)(methyl)carbamate, N-(tert-Butoxycarbonyl)-N-methyl-2-aminoethanol, N-Boc-N-methylethanolamine, N-Boc-N-methylglycinol, N-(tert-Butoxycarbonyl)-N-methylglycinol, (2-Hydroxyethyl)(methyl)carbamic acid tert-Butyl ester

B5685-1G B5685-5G
TCIAB5685-1GEA 22 EUR
TCIAB5685-1G TCIAB5685-5G
tert-Butyl (2-hydroxyethyl)(methyl)carbamate ≥98.0% (por GC)
tert-Butyl (2-hydroxyethyl)(methyl)carbamate
Formula: C₈H₁₇NO₃
MW: 175.23 g/mol
Pto. Ebullición: 84°C
Número MDL: MFCD03425857
Núm. CAS: 57561-39-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c3818c535713c6058abe993e7b0ebc48fd94d1c&vid=6c3818c535713c6058abe993e7b0ebc48fd94d1c&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR