(4-Chloro-2-methylphenoxy)acetic acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

C0206-25G C0206-500G
TCIAC0206-25GEA 32 EUR
TCIAC0206-25G TCIAC0206-500G
(4-Chloro-2-methylphenoxy)acetic acid ≥98.0% (by titrimetric analysis)
(4-Chloro-2-methylphenoxy)acetic acid
Formula: C₉H₉ClO₃
Número MDL: MFCD00004306
Núm. CAS: 94-74-6
EINECS: 202-360-6
Índice Merck: 14,05764

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 118.0 to 121.0 deg-C
Solubility in Methanol within almost transparency

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR