Cinchonidine sulphate dihydrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

C0349-25G
TCIAC0349-25GEA 75 EUR
TCIAC0349-25G
Cinchonidine sulphate dihydrate ≥98.0% (by titrimetric analysis)
Cinchonidine sulphate dihydrate
Formula: C₁₉H₂₂N₂O
Número MDL: MFCD00067565
Núm. CAS: 524-61-8
EINECS: 208-362-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water 6.5 to 7.2 %

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR