Carbonohidrazida ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

C0803-25G C0803-500G
TCIAC0803-25GEA 21 EUR
TCIAC0803-25G TCIAC0803-500G
Carbonohidrazida ≥98.0% (by titrimetric analysis)
Carbonohidrazida
Formula: CH₆N₄O
Temperatura de almacenaje: Nevera
Número MDL: MFCD00007591
Núm. CAS: 497-18-7
EINECS: 207-837-2
UN: 3077
ADR: 9,III
Índice Merck: 14,01804
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=729a8a8dffde7cbe7eed9482652dd3a91702d4f4&vid=729a8a8dffde7cbe7eed9482652dd3a91702d4f4&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Potassium iodate Method) min. 98.0 %
Melting point (Decomposition) 153.0 to 157.0 deg-C
Solubility in Water almost transparency

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR