Sodium hexanoate ≥99.0% (by titrimetric analysis)

Proveedor: TCI

C1187-25G
TCIAC1187-25GEA 168 EUR
TCIAC1187-25G
Sodium hexanoate ≥99.0% (by titrimetric analysis)
Sodium hexanoate
Formula: C₆H₁₁NaO₂
Número MDL: MFCD00059056
Núm. CAS: 10051-44-2
EINECS: 233-179-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f34807a2d78a82682ddaaeb525bac577c99b7a1d&vid=f34807a2d78a82682ddaaeb525bac577c99b7a1d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 % (after drying)
Drying loss max. 3.0 %
Sulfate to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR