N-(Carbobenzoxy)aminoacetonitrile ≥95.0% (by total nitrogen basis)

Proveedor: TCI
Atención

Sinónimos: Benzyl (cyanomethyl)carbamate, Cyanomethylcarbamic Acid Benzyl Ester, Benzyl Cyanomethylcarbamate, N-Cbz-aminoacetonitrile

C1439-25G C1439-5G
TCIAC1439-25GEA 610 EUR
TCIAC1439-25G TCIAC1439-5G
N-(Carbobenzoxy)aminoacetonitrile ≥95.0% (by total nitrogen basis)
N-(Carbobenzoxy)aminoacetonitrile
Formula: C₁₀H₁₀N₂O₂
MW: 190.20 g/mol
Número MDL: MFCD00033175
Núm. CAS: 3589-41-1

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (with Total Nitrogen) min. 95.0 %
Melting point 58.0 to 61.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR