(S)-(+)-Camptothecin ≥97.0% (by HPLC)

Proveedor: TCI
Peligro

C1495-1G C1495-100MG
TCIAC1495-1GEA 198 EUR
TCIAC1495-1G TCIAC1495-100MG
(S)-(+)-Camptothecin ≥97.0% (by HPLC)
(S)-(+)-Camptothecin
Formula: C₂₀H₁₆N₂O₄
Número MDL: MFCD00081076
Núm. CAS: 7689-03-4
UN: 1544
ADR: 6.1,III
Índice Merck: 14,01735

Order Now

Resultados test especificaciones

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Specific rotation [a]20/D +37.0 to +42.0 deg (C=0.4, CHCl3:MeOHl=8:2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR