Chloromethyl phenyl sulphone ≥98.0% (por GC)

Proveedor: TCI
Atención

Sinónimos: ((Chloromethyl)sulphonyl)benzene, 1-(Chloromethylsulphonyl)benzene, Phenyl Chloromethyl Sulphone

C1571-25G C1571-5G
TCIAC1571-25GEA 365 EUR
TCIAC1571-25G TCIAC1571-5G
Chloromethyl phenyl sulphone ≥98.0% (por GC)
Chloromethyl phenyl sulphone
Formula: C₇H₇ClO₂S
MW: 190.64 g/mol
Pto. Ebullición: 130°C
Número MDL: MFCD00007551
Núm. CAS: 7205-98-3
EINECS: 230-581-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR