Citronellic acid ≥95.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

C1708-25ML
TCIAC1708-25MLEA 132 EUR
TCIAC1708-25ML
Citronellic acid ≥95.0% (by titrimetric analysis)
Citronellic acid
Formula: C₁₀H₁₈O₂
Número MDL: MFCD00002728
Núm. CAS: 502-47-6
EINECS: 207-939-7
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9c0b73d709434d88dbc9c8f01b832b2b4eb8826&vid=f9c0b73d709434d88dbc9c8f01b832b2b4eb8826&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR