Ácido 4-Cianofenilborónico (contains varying amounts of Anhydride)

Proveedor: TCI
Peligro

Sinónimos: 4-Cyanobenzeneboronic acid, (4-Cyanophenyl)metaboric acid, Dihydroxy(4-cyanophenyl)borane

C1778-1G C1778-25G C1778-5G
TCIAC1778-1GEA 27 EUR
TCIAC1778-1G TCIAC1778-25G TCIAC1778-5G
Ácido 4-Cianofenilborónico (contains varying amounts of Anhydride)
Ácido 4-Cianofenilborónico
Formula: C₇H₆BNO₂
MW: 146.94 g/mol
Número MDL: MFCD01318968
Núm. CAS: 126747-14-6
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b7fdb659f89d71dd36c4fa263942498f19cee6a&vid=7b7fdb659f89d71dd36c4fa263942498f19cee6a&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 and le 114.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR