5-(Carbobenzoxyamino)valeric acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI

C2067-25G C2067-5G
TCIAC2067-25GEA 653 EUR
TCIAC2067-25G TCIAC2067-5G
5-(Carbobenzoxyamino)valeric acid ≥98.0% (by titrimetric analysis)
5-(Carbobenzoxyamino)valeric acid
Formula: C₁₃H₁₇NO₄
Número MDL: MFCD00188458
Núm. CAS: 23135-50-4

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 103.0 to 107.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR