2-(Chloromethyl)-3,4-dimethoxypyridine hydrochloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

C2073-25G C2073-5G
TCIAC2073-25GEA 157 EUR
TCIAC2073-25G TCIAC2073-5G
2-(Chloromethyl)-3,4-dimethoxypyridine hydrochloride ≥98.0% (by titrimetric analysis)
2-(Chloromethyl)-3,4-dimethoxypyridine hydrochloride
Formula: C₈H₁₀ClNO₂
Número MDL: MFCD02181083
Núm. CAS: 72830-09-2
EINECS: 416-440-5
UN: 3077
ADR: 9,III

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR