N-Carbobenzoxy-D-leucine dicyclohexylammonium salt ≥98.0% (by titrimetric analysis)

Proveedor: TCI

C2135-1G C2135-5G
TCIAC2135-1GEA 21 EUR
TCIAC2135-1G TCIAC2135-5G
N-Carbobenzoxy-D-leucine dicyclohexylammonium salt ≥98.0% (by titrimetric analysis)
N-Carbobenzoxy-D-leucine dicyclohexylammonium salt
Formula: C₁₄H₁₉NO₄
Número MDL: MFCD10566885
Núm. CAS: 7662-58-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 152.0 to 156.0 deg-C
Specific rotation [a]20/D +7.0 to +8.0 deg (C=3, MeOH)

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR