2'-Deoxyuridina ≥99.0% (by HPLC)

Proveedor: TCI

D0060-1G D0060-25G D0060-5G
TCIAD0060-1GEA 17 EUR
TCIAD0060-1G TCIAD0060-25G TCIAD0060-5G
2'-Deoxyuridina ≥99.0% (by HPLC)
2'-Deoxyuridina
Formula: C₉H₁₂N₂O₅
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00006527
Núm. CAS: 951-78-0
EINECS: 213-455-7
Índice Merck: 14,02910
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4286e77149b42e0bb559960e74940a0d86acf1a6&vid=4286e77149b42e0bb559960e74940a0d86acf1a6&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 164.0 to 168.0 deg-C
Specific rotation [a]20/D +51.0 to +53.0 deg (C=1, 1mol/L NaOH sol.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR