Dicloruro de 4-hidroxi-m-fenilendiamonio ≥97.0% (by total nitrogen and titration analysis)

Proveedor: TCI
Peligro

Sinónimos: 4-Hydroxy-1,3-Phenylenediammonium Dichloride, 2,4-Diaminophenol Dihydrochloride, Amidol

D0080-25G D0080-500G
TCIAD0080-25GEA 24 EUR
TCIAD0080-25G TCIAD0080-500G
Dicloruro de 4-hidroxi-m-fenilendiamonio ≥97.0% (by total nitrogen and titration analysis)
Dicloruro de 4-hidroxi-m-fenilendiamonio
Formula: C₆H₁₀Cl₂N₂O
MW: 197.06 g/mol
Pto. de fusión: 205°C
Número MDL: MFCD00012979
Núm. CAS: 137-09-7
EINECS: 205-279-4
UN: 2811
ADR: 6.1 III
Índice Merck: 14,02983
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f37baabacaa8312642a4ebad23c7200b8fad9ebc&vid=f37baabacaa8312642a4ebad23c7200b8fad9ebc&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Amber to Dark green powder to crystal
Purity (Argentmetric Titration) min. 97.0 % (calcd.on dried substance)
Purity (with Total Nitrogen) min. 98.0 % (calcd.on dried substance)
Drying loss max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR