Plata dietilditiocarbamato ≥98.0% (by titrimetric analysis) for As analysis

Proveedor: TCI
Atención

Sinónimos: Dietilditiocarbamato de plata, (Dietilditiocarbamato-S,S')plata

D0490-25G D0490-5G
TCIAD0490-25GEA 133 EUR
TCIAD0490-25G TCIAD0490-5G
Plata dietilditiocarbamato ≥98.0% (by titrimetric analysis) for As analysis
Plata dietilditiocarbamato
Formula: (C₂H₅)₂NCSSAg
MW: 256.14 g/mol
Número MDL: MFCD00004929
Núm. CAS: 1470-61-7
EINECS: 216-003-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e92e256b5eb38b21bd24055a08e0e16f4e169cf&vid=bef3328062f5aab5bbd0782035bdf2bede2d9562&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow powder to crystal
Purity (NH4SCN method) min. 98.0 %
Solubility in Pyridine almost transparency
Suitability for arsenic-analysis to pass test

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR