Trisodium 2-(4-Sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonate (SPADNS) ≥95.0% (by HPLC)

Proveedor: TCI
Atención

D0604-25G D0604-5G
TCIAD0604-25GEA 200 EUR
TCIAD0604-25G TCIAD0604-5G
Trisodium 2-(4-Sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonate (SPADNS) ≥95.0% (by HPLC)
Trisodium 2-(4-Sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonate (SPADNS)
Formula: C₁₆H₁₂N₂O₁₁S₃
Número MDL: MFCD00003952
Núm. CAS: 23647-14-5
EINECS: 245-803-9

Order Now

Resultados test especificaciones

Appearance Orange to Brown to Dark red powder to crystal
Purity (HPLC) min. 95.0 area%
Water 3.0 to 14.0 %
Sensitiveness to pass test

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR