meso-ɑ,β-Di(4-pyridyl) glycol ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

D0936-5G
TCIAD0936-5GEA 130 EUR
TCIAD0936-5G
meso-ɑ,β-Di(4-pyridyl) glycol ≥98.0% (by HPLC, titration analysis)
meso-alpha,beta-Di(4-pyridyl) glycol
Formula: C₁₂H₁₂N₂O₂
Número MDL: MFCD00023627
Núm. CAS: 4972-49-0

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR