trans-3,4-Dimethyl-2-pentene ≥98.0% (por GC)

Proveedor: TCI

D1256-5ML
TCIAD1256-5MLEA 235 EUR
TCIAD1256-5ML
trans-3,4-Dimethyl-2-pentene ≥98.0% (por GC)
trans-3,4-Dimethyl-2-pentene

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR