9,10-Dicyanoanthracene ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Peligro

D1656-1G D1656-5G
TCIAD1656-1GEA 62 EUR
TCIAD1656-1G TCIAD1656-5G
9,10-Dicyanoanthracene ≥98.0% (by HPLC, total nitrogen)
9,10-Dicyanoanthracene
Formula: C₁₆H₈N₂
Número MDL: MFCD00041733
Núm. CAS: 1217-45-4
UN: 3439
ADR: 6.1,III

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR