1,1-Diphenyl-4-(piperidin-1-yl)butan-1-ol hydrochloride ≥99.0% (by titrimetric analysis)

Proveedor: TCI
Atención

D2062-25G
TCIAD2062-25GEA 65 EUR
TCIAD2062-25G
1,1-Diphenyl-4-(piperidin-1-yl)butan-1-ol hydrochloride ≥99.0% (by titrimetric analysis)
1,1-Diphenyl-4-(piperidin-1-yl)butan-1-ol hydrochloride
Formula: C₂₁H₂₈ClNO
Número MDL: MFCD00151479
Núm. CAS: 3254-89-5
EINECS: 221-850-0
Índice Merck: 14,03311

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR