5-Fluoro-2'-deoxyuridina (Floxuridina) ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

D2235-1G D2235-100MG D2235-500MG
TCIAD2235-1GEA 180 EUR
TCIAD2235-1G TCIAD2235-100MG TCIAD2235-500MG
5-Fluoro-2'-deoxyuridina (Floxuridina) ≥98.0% (by HPLC, titration analysis)
5-Fluoro-2'-deoxyuridina (Floxuridina)
Formula: C₉H₁₁FN₂O₅
Número MDL: MFCD00006530
Núm. CAS: 50-91-9
EINECS: 200-072-5
UN: 2811
ADR: 6.1,III
Índice Merck: 14,04112
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=757aa2698697c2d0d76220337d62c82dd3011395&vid=757aa2698697c2d0d76220337d62c82dd3011395&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 171.0 to 175.0 deg-C
Solubility in Acetonitrile very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR