4'-(Dimethylamino)benzylidene-4-nitroaniline

Proveedor: TCI

D2264-5G
TCIAD2264-5GEA 81 EUR
TCIAD2264-5G
4'-(Dimethylamino)benzylidene-4-nitroaniline
4'-(Dimethylamino)benzylidene-4-nitroaniline
Formula: C₁₅H₁₅N₃O₂
Número MDL: MFCD00086550
Núm. CAS: 896-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR