1,6-Difenil-1,6-butanodiona ≥98.0% (por GC)

Proveedor: TCI

D2773-25G D2773-5G
TCIAD2773-25GEA 671 EUR
TCIAD2773-25G TCIAD2773-5G
1,6-Difenil-1,6-butanodiona ≥98.0% (por GC)
1,6-Difenil-1,6-butanodiona
Formula: C₁₈H₁₈O₂
Número MDL: MFCD00004761
Núm. CAS: 3375-38-0
EINECS: 222-166-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=701893a830daf2dbf61834819162e38c1557f5b1&vid=701893a830daf2dbf61834819162e38c1557f5b1&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C
Solubility in hot Toluene almost transparency

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR