N,N'-Dimethylquinacridone 96,0 HPLC_ASSAY_METHOD, purified_sublimation

Proveedor: TCI

Sinónimos: DMQA, 5,12-Dihydro-5,12-dimethylquino[2,3-b]acridine-7,14-dione

D3227-1G
TCIAD3227-1GEA 103 EUR
TCIAD3227-1G
N,N'-Dimethylquinacridone 96,0 HPLC_ASSAY_METHOD, purified_sublimation
N,N'-Dimethylquinacridone
Formula: C₂₂H₁₆N₂O₂
MW: 340.38 g/mol
Número MDL: MFCD00683078
Núm. CAS: 19205-19-7

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 96.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR