Diundecyl phthalate (mixture of branched chain isomers) ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Phthalic Acid Diundecyl Ester

D3248-500G
TCIAD3248-500GEA 33 EUR
TCIAD3248-500G
Diundecyl phthalate (mixture of branched chain isomers) ≥98.0% (by titrimetric analysis)
Diundecyl phthalate (mixture of branched chain isomers)
Formula: C₃₀H₅₀O₄
MW: 474.73 g/mol
Pto. Ebullición: 267°C
Pto. de inflamación: 239°C
Núm. CAS: 96507-86-7
EINECS: 306-165-8

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Purity (Ester Value) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR