3,5-Dimethyl-1-adamantanamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI
Atención

D3608-25G D3608-5G
TCIAD3608-25GEA 201 EUR
TCIAD3608-25G TCIAD3608-5G
3,5-Dimethyl-1-adamantanamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
3,5-Dimethyl-1-adamantanamine hydrochloride
Formula: C₁₂H₂₁N
Pto. de fusión: 292°C
Número MDL: MFCD00214336
Núm. CAS: 41100-52-1
EINECS: 255-219-6
Índice Merck: 14,05806

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR