3,4-Dihydroxy-5-nitrobenzaldehyde ≥98.0% (por GC)

Proveedor: TCI
Peligro

D3645-25G
TCIAD3645-25GEA 112 EUR
TCIAD3645-25G
3,4-Dihydroxy-5-nitrobenzaldehyde ≥98.0% (por GC)
3,4-Dihydroxy-5-nitrobenzaldehyde
Formula: C₇H₅NO₅
Número MDL: MFCD00871897
Núm. CAS: 116313-85-0
EINECS: 441-810-8

Order Now

Resultados test especificaciones

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 145.0 to 149.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR