o-Dianisidina ≥95.0% (by titrimetric analysis) for biochemical research

Proveedor: TCI
Peligro

Sinónimos: 3,3'-Dimetoxibencidina

D3864-1G D3864-5G
TCIAD3864-1GEA 16 EUR
TCIAD3864-1G TCIAD3864-5G
o-Dianisidina ≥95.0% (by titrimetric analysis) for biochemical research
o-Dianisidina
Formula: C₁₄H₁₆N₂O₂
MW: 244.29 g/mol
Número MDL: MFCD00008372
Núm. CAS: 119-90-4
EINECS: 204-355-4
UN: 2811
ADR: 6.1 III
Índice Merck: 14,02991
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06f8fb67cf7cc962c48a5a8dbbca316052008850&vid=4a65e0285ab104a27fbab9e400bd0a912b3c9622&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 136.0 to 140.0 deg-C
Hydrogendioxide detection test (Peroxidase method) to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR