3,3'-Diethyloxadicarbocyanine iodide (DODC) ≥98.0% (by total nitrogen basis)

Proveedor: TCI
Atención

D4457-200MG
TCIAD4457-200MGEA 49 EUR
TCIAD4457-200MG
3,3'-Diethyloxadicarbocyanine iodide (DODC) ≥98.0% (by total nitrogen basis)
3,3'-Diethyloxadicarbocyanine iodide (DODC)
Formula: C₂₃H₂₃IN₂O₂
Número MDL: MFCD00011953
Núm. CAS: 14806-50-9
EINECS: 238-873-7

Order Now

Resultados test especificaciones

Appearance Blue to Dark blue powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Absorbance (E1%1cm) min. 4500 (MeOH, 576.0 to 582.0 nm)
Lambda max. 576.0 to 582.0 nm (MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR