Dextrina

Proveedor: TCI

D4657-100G D4657-500G
TCIAD4657-100GEA 21 EUR
TCIAD4657-100G TCIAD4657-500G
Dextrina
Dextrina
Formula: (C₆H₁₂O₆)n
Número MDL: MFCD00081554
Núm. CAS: 9004-53-9
EINECS: 232-675-4
Índice Merck: 14,02953

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Drying loss max. 10.0 %
Ignition residue (Sulfate) max. 0.5 %
Reducing suger max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR