2-Dicyclohexylphosphino-2',6'-diisopropoxybiphenyl ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

D5037-1G D5037-5G
TCIAD5037-1GEA 107 EUR
TCIAD5037-1G TCIAD5037-5G
2-Dicyclohexylphosphino-2',6'-diisopropoxybiphenyl ≥98.0% (by HPLC, titration analysis)
2-Dicyclohexylphosphino-2',6'-diisopropoxybiphenyl
Formula: C₃₀H₄₃O₂P
Número MDL: MFCD06798294
Núm. CAS: 787618-22-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=342c81f29df38a09003bee6cb63d45268d158fa9&vid=342c81f29df38a09003bee6cb63d45268d158fa9&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 123.0 to 127.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR