9,10-Di(2-naphthyl)anthracene ≥99.0% (by HPLC), purified by sublimation

Proveedor: TCI

D5066-200MG
TCIAD5066-200MGEA 58 EUR
TCIAD5066-200MG
9,10-Di(2-naphthyl)anthracene ≥99.0% (by HPLC), purified by sublimation
9,10-Di(2-naphthyl)anthracene
Formula: C₃₄H₂₂
Pto. de fusión: 357°C
Número MDL: MFCD00028944
Núm. CAS: 122648-99-1

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 99.0 area%

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR