3,5-Dimethyl-4-propoxyphenylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: (3,5-Dimethyl-4-propoxyphenyl)boronic acid

D5080-1G D5080-5G
TCIAD5080-1GEA 20 EUR
TCIAD5080-1G TCIAD5080-5G
3,5-Dimethyl-4-propoxyphenylboronic acid (contains varying amounts of Anhydride)
3,5-Dimethyl-4-propoxyphenylboronic acid
Formula: C₁₁H₁₇BO₃
Número MDL: MFCD05865178
Núm. CAS: 357611-51-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a600ecf65ccf9b19cd87d7a17accf1217add32db&vid=a600ecf65ccf9b19cd87d7a17accf1217add32db&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR